Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Staphylococcus Glu-C Recombinant Protein, PeproTech®

Catalog No. 450461MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
250 μg
50 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-46-1MG 1 mg
45046250UG 250 μg
45046500UG 500 μg
450-46-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 450-46-1MG Supplier Gibco™ Supplier No. 450461MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Staphylococcus Glu-C is a 28.8 kDa protease consisting of 266 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to, specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, as they are frequently used in the analysis and production of proteins. Glu-C cleaves at the Carboxyl side of E (can also cleave D under certain conditions).
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 66839245
Molecular Weight (g/mol) 28.8 kDa
Name Staphylococcus Glu-C
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Common Name Glu-C
Gene Symbol sspA
Biological Activity Cleaves at the Carboxyl side of E (can also cleave D under certain conditions).
Conjugate Unconjugated
Recombinant Recombinant
Sequence LPNNDRHQIT DTTNGHYAPV TYIQVEAPTG TFIASGVVVG KDTLLTNKHV VDATHGDPHA LKAFPSAINQ DNYPNGGFTA EQITKYSGEG DLAIVKFSPN EQNKHIGEVV KPATMSNNAE TQVNQNITVT GYPGDKPVAT MWESKGKITY LKGEAMQYDL STTGGNSGSP VFNEKNEVIG IHWGGVPNEF NGAVFINENV RNFLKQNIED IHFANDDQPN NPDNPDNPNN PDNPNNPDEP NNPDNPNNPD NPDNGDNNNS DNPDAA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.