Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch Indicator, 0.5% (w/v), with Chloroform preservative, Ricca Chemical
SDP

Catalog No. 800232 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
800232 1 L Natural Poly Bottle
800216 500 mL Natural Poly Bottle
80021 4 L Natural Poly Bottle
80025 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 800232 Supplier Ricca Chemical Company Supplier No. 800232
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Starch Indicator
Name Note With Chloroform Preservative
CAS 7732-18-5, 9005-84-9, 67-66-3
CAS Min % 98.0, 0.49, 0.1
CAS Max % 100.0, 0.51, 0.4
Concentration 0.5% (w/v)
Percent Purity 0.5% W/V
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Colorless
pH 5 to 7
Quantity 1 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.