Learn More
Starch Indicator, Preserved with Salicylic Acid (Mercury Free), Certified, 0.5%, LabChem™
Specifications
Specifications
| Chemical Name or Material | Starch Indicator |
| Name Note | Preserved with Salicylic Acid (Mercury Free) |
| CAS Max % | 99.4, 0.5, 0.1 |
| CAS | 9005-25-8, 69-72-7 |
| Solubility Information | Soluble in water |
| Identification | Passes Test |
| Percent Purity | 0.5% |
| Grade | Certified |
| Packaging | Cubitainer™ |
| Specific Gravity | 1g/mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.