Learn More
Starch Indicator Solution, 1.0% w/v, AquaPhoenix Scientific™
Shop All AquaPhoenix Scientific Products

Specifications
Specifications
| CAS | 9005-84-9, 69-72-7, 7732-18-5 |
| Concentration | 1.0% w/v |
| Packaging | Poly Bottle, Purple Cap |
| Quantity | 500 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.