Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch, Soluble, ACS Reagent, Reagents

Catalog No. 01337652
Change view
Click to view available options
Quantity:
125 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-652 500 g
01-337-654 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-652 Supplier Reagents Holdings Llc Supplier No. C1332000500A5
Add to Cart
Add to Cart

Starch, Soluble, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 9005-84-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
InChI Key GUBGYTABKSRVRQ-ASMJPISFSA-N
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Boiling Range 668°C
Color White
Boiling Point 668°C
Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1.5
Grade ACS
Physical Form Solid
Chemical Name or Material Starch, Soluble, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.