Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch, Soluble, Laboratory Grade, Innovating Science™
GREENER_CHOICESCI_ED

Catalog No. S000209 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000209 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000209 Supplier Aldon Corporation Supplier No. IS28205
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9005-84-9

Specifications

Packaging HDPE
Quantity 100 g
Percent Purity 100%
Grade Laboratory
Chemical Name or Material Starch
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.