Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STARD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1562 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1562 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1562 Supplier Invitrogen™ Supplier No. PA1562
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA1-562 detects MLN64 from rat tissues. PA1-562 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~53 kDa protein representing MLN64 from rat adrenal gland tissue extracts. This antibody also detects an ~25 kDa protein from rat adrenal gland which may correspond to degradation product. PA1-562 antigen is recombinant MLN64 protein.

The steroidogenic acute regulatory (StAR) protein facilitates the movement of cholesterol from the outer to inner mitochondrial membrane in adrenal and gonadal cells, fostering steroid biosynthesis. MLN64 is a 445-amino acid protein of unknown function. When 218 amino-terminal residues of MLN64 are deleted, the resulting N-218 MLN64 has 37% amino acid identity with StAR and 50% of StAR's steroidogenic activity in transfected cells. Bacterially expressed N-218 MLN64 exerts StAR-like activity to promote the transfer of cholesterol from the outer to inner mitochondrial membrane in vitro. The presence of a protease-resistant domain and a protease-sensitive carboxy-terminal domain in N-218 MLN64 is similar to the organization of StAR. However, as MLN64 never enters the mitochondria, the protease-resistant domain of MLN64 cannot be a mitochondrial pause-transfer sequence, as has been proposed for StAR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STARD3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene STARD3
Gene Accession No. 0
Gene Alias CAB1; Es64; metastatic lymph node gene 64 protein; metastatic lymph node protein 64; MLN 64; MLN64; Protein CAB1; Protein ES 64; protein MLN 64; StAR related lipid transfer domain containing 3; STARD3; StAR-related lipid transfer (START) domain containing 3; StAR-related lipid transfer domain containing 3; stAR-related lipid transfer protein 3; START domain containing 3; START domain-containing protein 3; steroidogenic acute regulatory protein related
Gene Symbols STARD3
Host Species Rabbit
Immunogen Recombinant MLN64 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363675
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.