Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STARD3NL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116687 Supplier Invitrogen™ Supplier No. PA5116687
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Jurkat, Mouse spleen, Rat spleen Immunogen sequence: RLLIVQDASE RAALIPGGLS DGQFYSPPES EAGSEEAEEK QDSEKPLLEL.

This gene encodes a late-endosomal protein that contains a conserved MENTAL (MLN64 N-terminal) domain. The encoded protein binds cholesterol molecules and may play a role in endosomal cholesterol transport through interactions with metastatic lymph node protein 64 (MLN64).
TRUSTED_SUSTAINABILITY

Specifications

Antigen STARD3NL
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Stard3nl
Gene Accession No. O95772, Q9DCI3
Gene Alias 0610035N01Rik; 6530409L22Rik; AW124774; MENTHO; MGC3251; MLN64 N-terminal domain homolog; MLN64 N-terminal homolog; STARD3 N-terminal like; STARD3 N-terminal-like protein; STARD3NL; UNQ855/PRO1864
Gene Symbols Stard3nl
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 185-234 of human STARD3NL (NP_1144051).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291182, 76205, 83930
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.