Learn More
METTLER TOLEDO™ STARe Software Option DSC Evaluation
Catalog No. 01915203 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
30421563
Catalog No. For Use With (Equipment)
01-915-203 30421563
1 options
Catalog No. 01-915-203 Supplier METTLER TOLEDO™ Supplier No. 119457
Add to Cart
Add to Cart
Request a Quote
DSC evaluation provides DSC users with additional evaluation capabilities. Included are the routines for the determination of glass transitions, the crystallinity, the content according to DIN, ASTM and IEC standard methods.
  • Content, crystallinity and enthalpy- this software option includes content, crystallinity and enthalpy routines
  • Conversion routine- the SW option contains a conversion routine, the basis for most kinetics measurements
  • Glass transition determination- the SW option allows the determination of the glass transition temperature with or without a relaxation peak
For use with 30421563, 30319177, 30421564 and 30319178
TRUSTED_SUSTAINABILITY
Specifications
Description STARe software
For Use With (Application) METTLER TOLEDO DSC
For Use With (Equipment) 30421563