Learn More
METTLER TOLEDO™ STARe Software Option TGA Evaluation
Catalog No. 01915202 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
30319250
Catalog No. For Use With (Equipment)
01-915-202 30319250
1 options
Catalog No. 01-915-202 Supplier METTLER TOLEDO™ Supplier No. 119459
Add to Cart
Add to Cart
Request a Quote
This STARe TGA Evaluation Software option provides additional evaluation capabilities such as content determination or conversion calculation. This kind of data is required for kinetics applications.
  • Content determination - the evaluation possibilities allow the determination of content - percent, stoichiometric, empirical - of the investigated substance
  • Conversion routine- the SW option contains a conversion routine, the basis for most kinetics measurements
For use with 30319250, 30319252, 30421562, 30319254 and 30293533
TRUSTED_SUSTAINABILITY
Specifications
Description STARe software
For Use With (Application) METTLER TOLEDO TGA
For Use With (Equipment) 30319250