Learn More
Heidolph™ StarFish™ Distribution Manifolds
SDP
Catalog No. 05404063 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
Quick Release Female
Quick Release Male
Size:
6.4 mm
9.5 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Size
05-404-063 Quick Release Male 6.4 mm
05-404-065 Quick Release Female 9.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-404-063 Supplier Heidolph™ Supplier No. 015890360
Add to Cart
Add to Cart
Heidolph™ StarFish™ Distribution Manifolds available in two different types, includes shut-off
  • For use with the StarFish™ Work Station
  • Allows for the connection of either coolants, or gas/vacuum
Specifications
Description RA Barbed
For Use With (Equipment) StarFish Work Station
For Use With (Application) Allows for the Connection of Coolant
Includes Shut-off (3.2 mm ID)
Quantity 5/Pk.
Size 6.4 mm
Type Quick Release Male