Learn More
Starplex™ Scientific Kangapack™ Specimen Containers with O-Ring Cap
Catalog No. 1490555 Shop All Starplex Scientific Products
Change view
Click to view available options
Capacity (Metric):
20 mL
90 mL
Product Type:
Primary Container
Secondary Container
Closure Type:
DisCap
O-Ring Cap
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Product Type Closure Type
14-905-55 90 mL Primary Container O-Ring Cap
14-905-58 20 mL Secondary Container DisCap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-905-55 Supplier Starplex™ Scientific Supplier No. K9021WNL
This item is not returnable. View return policy
Patented design nests secondary container inside the primary container for convenient handling of split specimens
  • Minimum fill line on primary and secondary containers
  • O-ring cap design grips top and sides of container for seal
Specifications
Product Type Primary Container
Material Plastic
Capacity (Metric) 90 mL
Closure Type O-Ring Cap