Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific Sanicult™ Hygiene Monitoring Test Kit

Catalog No. 14959200 Shop All Starplex Scientific Products
Change view
Click to view available options
Quantity:
100/Bag
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-959-200 100/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-959-200 Supplier Starplex™ Scientific Supplier No. B30US
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A sterile buffered solution for sampling environmental surfaces. Can be used for qualitative and quantitative procedures. A Peptone Broth is used for pre-enrichment of Salmonella from food.Designed for industrial applications.

  • Includes swab and tube containing sterile diluent solution for testing surfaces, viscous liquids, semi-solids and powders.
  • Designed for industrial application (food and beverage, engineering and allied industries, petrochemical, textile, water testing).
  • Simple and time-saving

 

Specifications

For Use With (Application) Industrial applications
Quantity 100/Bag
Storage Requirements 4°C to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.