Description
- Break- and leak-resistant for safety during harsh transport conditions or pneumatic tube systems
- Clarified polypropylene cups provide easy visual analysis of sample
- Full-circle thread design prevents cross-threading
- Graduated markings allow quick measurement of sample
- Labeling provides ample space for patient information
- Ridges on cap provide a secure grip for safe handling
Specifications
Specifications
| Graduated | Yes |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Capacity (Metric) | 192 mL |
| Closure Color | Yellow |
| Closure Type | Spout Cap |