Learn More
TARGETMOL CHEMICALS INC STAT3-IN-B9 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T28865
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. STAT3-IN-B9 (B9) is an inhibitor of abnormal STAT3 activation and inhibits the proliferation of tumor cells including MDA-MB-468 MDA-MB-231 and DU145. purity: 98%
Catalog No. 50-258-0486
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.