Learn More
Stat4 Mouse anti-Human, Clone: OTI2A2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500005

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The STAT 4 protein is a transcription factor activated by hormone and cytokines. STAT 4 is most similar to STAT 1 (52%) and STAT 3 (47%). Functionally, STAT 4 is similar to other STAT family members in that it can be tyrosine phosphorylated by JAK 1 or JAK 2. STAT 4 forms homodimers and heterodimers with related STAT family members. Tyrosine phosphorylated STAT 4 can bind the interferon g activated site (GAS), and serine phosphorylation is also required for maximal transcriptional activity. STAT 4 expression is restricted to the thymus, spleen, and testis.Specifications
Stat4 | |
Monoclonal | |
Unconjugated | |
STAT4 | |
SLEB11 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6775 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2A2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q14765 | |
STAT4 | |
Recombit fragment expressed in E.coli corresponding to amino acids 633-652 of human Stat4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.