Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAT5 alpha/beta Monoclonal Antibody (ST5-8F7)
GREENER_CHOICE

Catalog No. 335900
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-590-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33-590-0 Supplier Invitrogen™ Supplier No. 335900
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

STAT5 alpha/beta Monoclonal Antibody for ELISA, ICC/IF, Western Blot

The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor-associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein is activated by, and mediates the responses of, many cell ligands, such as IL-2, IL-3, IL-7, GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of this protein in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of this gene is found to induce the expression of BCL2L1/ BCL-X(L), which suggests the antiapoptotic function of this gene in cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAT5 alpha/beta
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone ST5-8F7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene STAT5A
Gene Accession No. P42229, P42230, P42232, P51692, P52632, Q62771
Gene Alias AA959963; Mammary gland factor; mammary gland factor STAT5A; mammary gland factor STAT5B; mammary gland factor; transcription factor; MGF; Mpf; signal transducer and activator of transcription 5; Signal transducer and activator of transcription 5a; Signal transducer and activator of transcription 5B; STA5B; STAT; STAT5; Stat5a; Stat5b; transcription factor STAT5B
Gene Symbols STAT5A, STAT5B
Host Species Mouse
Immunogen A peptide corresponding to a conserved amino acid sequence found near the carboxy-terminus of murine STAT5. This peptide encompasses the conserved C-terminal tyrosine phosphorylation site (Y-694) of murine STAT5.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20850, 20851, 24918, 25126, 6776, 6777
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.