Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Stathmin 1 Recombinant Superclonal™ Antibody (4 HCLC)

Catalog No. 711090
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-109-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-109-0 Supplier Invitrogen™ Supplier No. 711090
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with Monkey, Dog, Cat and Sheep. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

Stathmin 1 belongs to the stathmin family of genes. It encodes a ubiquitous cytosolic phosphoprotein proposed to function as an intracellular relay integrating regulatory signals of the cellular environment. Stathmin 1 is involved in the regulation of the microtubule filament system by destabilizing microtubules. It prevents assembly and promotes disassembly of microtubules. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stathmin 1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Clone 4 HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene Stmn1
Gene Accession No. P13668, P16949, P54227
Gene Alias 19k; C1orf215; I79_017954; Lag; LAP18; leukemia associated phosphoprotein p18; leukemia-associated cytosolic phosphoprotein stathmin; leukemia-associated gene protein; Leukemia-associated phosphoprotein p18; metablastin; Oncoprotein 18; Op18; OTTHUMP00000008529; OTTHUMP00000008531; P18; P19; phosphoprotein 19; phosphoprotein p19; Pig; pp17; Pp18; Pp19; Pr22; Pr22 protein; Prosolin; Protein Pr22; RP1-125I3.5; SMN; Stathmin; stathmin 1; stathmin 1 variant; stathmin 1/oncoprotein 18; Stmn1; STN1; testicular tissue protein Li 189; transmembrane protein C1orf215
Gene Symbols Stmn1
Host Species Rabbit
Immunogen Peptides corresponding to Human STMN1 (aa 25-37, 5-14).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16765, 29332, 3925
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.