Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Stationary Vehicle Placards, FLAMMABLE GAS 2

Catalog No. 19111343 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Fiberglass (B-120)
Polycoated Tagstock (B-101)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-111-343 Fiberglass (B-120)
19-111-346 Polycoated Tagstock (B-101)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-111-343 Supplier Brady™ Supplier No. 63438
Add to Cart
Add to Cart

Indicate delivery vehicles carrying dangerous or hazardous materials. Brady™ Stationary Vehicle Placards are available in a variety of materials.

  • Available in two different materials Polycoated Tagstock and Premium Fiberglass

Specifications

Adhesive Type Non-adhesive
Color White, Red
Material Fiberglass (B-120)
Product Type Vehicle Placard
Display Text FLAMMABLE GAS 2
For Use With (Application) Material Handling, Shipping and Receiving Signs and Labels
Quantity 1 Ea.
Shape Diamond
Size (English) 10.75 x 10.75 in.
Thickness (English) 0.0984 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.