Learn More
Essendant STATIONUSB-C DOCKSD2BK
Supplier:  Essendant KMWK38260NA
Encompass_Preferred
Ideal for hot-desking hoteling home office and educational environments requiring a single monitor solution Kensington s SD2000P USB-C Single 4K Nano Dock with 60 W Power Delivery is Thunderbolt 3 compatible and provides users with a simple connection to a desktop setup The SD2000P allows users to choose between HDMI or DPplus plus video ports to accommodate different monitor setups connect to their favorite USB accessories using two USB 3 1 Gen1 ports and use the USB-C 3 2 Gen1 connection to transfer up to 5 Gbps of data while powering their device For Use With- USB-C Devices Operating System Compatibility- Android OS 9 and Later Chrome OS 44 and Later iPad OS 13 4 and Later Mac OS 10 13 and Later Windows 7 and Later Connector/Port/Interface- USB-C Batteries Required- No
Catalog No. 50-000-34266
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.