Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desco™ Statshield™ Moisture Barrier Bag
SDP

Catalog No. 18688068
Encompass
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W):
10 x 8 in.
20 x 10 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18-688-068 10 x 8 in.
18-688-073 20 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-688-068 Supplier Desco™ Supplier No. 13812
Add to Cart
Add to Cart

Barrier Bag designed for electrostatic discharge (ESD) shielding protection.

  • Film laminate metal
  • Meets ANSI/ESD S20.20, ANSI/ESD S541, EIA 541, JESD625-A, and EIA 583
  • Designed for dry packing of moisture sensitive devices and to protect contents from damage due to ESD events or EMI
  • < 10 nJ Energy Penetration Tested per ANSI/ESD STM11.31
  • Protects contents from ESD charges and discharges
  • Provides physical protection for contents
  • Dissipative Inner and Outer Surfaces Rtt < 1 x 1011 ohms per ANSI/ESD STM11.11 allows bag to be grounded when placed on a grounded surface
  • Printed with ESD Protective Symbol and lot code information
  • Lead-free RoHS compliant

Specifications

Color Silver
For Use With (Application) ESD sensitive devices
Material Aluminized Polyester
Quantity 100/Pk.
Thickness (English) 3.50 mil
Dimensions (L x W) 10 x 8 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.