Learn More
HemoCue America StatSpin™ Blood Separation Prepared Microtubes
Catalog No. 22269523
Change view
Click to view available options
Capacity (Metric):
1.3 mL
Catalog No. Capacity (Metric)
22-269-523 1.3 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-269-523 Supplier HemoCue America Supplier No. TP1U
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
•    Serum (red top), plasma (green top), or gel tubes (clear top)
•    Separations in only 30 to 90 seconds
•    Fit RT 12 tube rotor, Cat. No. 22-269-258
•    For use with StatSpin VT Veterinary Multipurpose Centrifuges 22-269-520, -521
•    May also be used with the StatSpin MP (22-314-934), RP (22-314-935) and CritSpin™ (22-269-291, -292, -294, 22-305-276) centrifuges
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1.3 mL
Material Plastic
Closure Type Snap Cap