Learn More
Selleck Chemical LLC Stattic 10mM/1mL 19983-44-9
SDP
Supplier:  Selleck Chemical LLC S702410MM/1ML
Encompass_Preferred
Stattic, the first nonpeptidic small molecule, potently inhibits STAT3 activation and nuclear translocation with IC50 of 5.1 ?M in cell-free assays, highly selectivity over STAT1. Stattic induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4327
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.