Learn More
Enzo Life Sciences Stattic (50mg). CAS: 19983-44-9
SDP
Supplier:  Enzo Life Sciences BMLEI3680050
Encompass_Preferred
Small molecule STAT3-specific inhibitor which blocks activation, dimerization and nuclear translocation via binding to the STAT3 SH2 domain. It is a very useful tool for exploring STAT3-mediated signaling pathways. It blocks LPS-mediated STAT3 tyrosine phosphorylation which leads to inhibition of LPS-mediated IL-1β and IL-6 production. In primary cultures of rat neural stem cells it blocks IL-15-induced differentiation. Alternative name: 6-Nitro[b]thiophene-1,1-dioxide. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25 mg/ml), 100% ethanol (10 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0752
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.