Learn More
AG Scientific Inc Staurosporine 5 MG
Supplier:  AG Scientific Inc S10165MG
Encompass_Preferred
Staurosporine is a cell-permeable protein kinase inhibitor that acts by blocking interaction with the ATP binding site. It mainly inhibits protein kinase C (PKC) but most other kinases, including tyrosine kinases. It has some antifungal properties.It has been reported that Staurosporine promotes apoptosis in various cell types by triggering both the morphological changes and intranucleosomal DNA fragmentation and by caspase-dependent and -independent ways. In addition, it has been reported that the kinase inhibitor is related to signaling pathway.
Catalog No. 50-226-8380
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.