Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Staurosporine, 98%

Catalog No. AC328532500
Change view
Click to view available options
Quantity:
1 mg
250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328532500 250 μg
AC328530010 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC328532500 Supplier Thermo Scientific Chemicals Supplier No. 328532500
  • $523.00
    $423.65 / Each of 1

    Save $99.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Staurosporine (staurosporin, CAS # 62996-74-1) is a bacterial alkyloid isolated from Streptomyces staurosporeus.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Staurosporine is an indolocarbazole alkaloid capable of permeating cell membranes
  • Staurosporine has anti-cancer properties that can induce apoptosis
  • Staurosporine is a broad inhibitor of protein kinases, preventing growth and division cycles

Application

  • Staurosporine is used for research on acute myeloid leukemia
  • It is suitable for research applications including cell culture, in vivo studies, and biochemistry
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62996-74-1
Molecular Formula C28H26N4O3
Molecular Weight (g/mol) 466.5
MDL Number MFCD00077402
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
Synonym staurosporine, kinome_3629
PubChem CID 49831000
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC

Specifications

Melting Point 270.0°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 μg
Merck Index 15,8928
Solubility Information Soluble in DMSO, ethyl acetate or dimethyl formamide. Insoluble in water.
Formula Weight 466.5
Percent Purity 98%
Chemical Name or Material Staurosporine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.