CAS RN 62996-74-1
Staurosporine, 99+%, Thermo Scientific Chemicals
CAS: 62996-74-1 Molecular Formula: C28H26N4O3 Molecular Weight (g/mol): 466.541 MDL Number: MFCD00077402 InChI Key: HKSZLNNOFSGOKW-ZYSRIHRCSA-N Synonym: Antibiotic AM-2282 PubChem CID: 49831000 SMILES: CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC
Thermo Scientific Chemicals Staurosporine, 1 mg/ml in DMSO, sterile-filtered
CAS: 62996-74-1 Molecular Formula: C28H26N4O3 Molecular Weight (g/mol): 466.541 MDL Number: MFCD00077402 InChI Key: HKSZLNNOFSGOKW-ZYSRIHRCSA-N Synonym: staurosporine,kinome_3629 PubChem CID: 49831000 SMILES: CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC
Thermo Scientific Chemicals Staurosporine, 98%
CAS: 62996-74-1 Molecular Formula: C28H26N4O3 Molecular Weight (g/mol): 466.5 MDL Number: MFCD00077402 InChI Key: HKSZLNNOFSGOKW-ZYSRIHRCSA-N Synonym: staurosporine,kinome_3629 PubChem CID: 49831000 SMILES: CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC
MilliporeSigma™ Calbiochem™ Staurosporine, Streptomyces sp.
Staurosporine, CAS 62996-74-, is a cell-permeable, potent, reversible, ATP-competitive inhibitor of protein kinases (IC50 = 7, 20, 1.3, 0.7, & 8.5 nM for PKA, CAMK, MLCK, PKC, & PKG, respectively).
Staurosporine, MP Biomedicals™
CAS: 62996-74-1 Molecular Formula: C28H26N4O3 Molecular Weight (g/mol): 466.541 InChI Key: HKSZLNNOFSGOKW-ZYSRIHRCSA-N Synonym: Staurosporin, Antibiotic AM-2282 PubChem CID: 49831000 SMILES: CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC
Staurosporine, MP Biomedicals™
CAS: 62996-74-1 Molecular Formula: C28H26N4O3 Molecular Weight (g/mol): 466.541 InChI Key: HKSZLNNOFSGOKW-ZYSRIHRCSA-N Synonym: staurosporine,kinome_3629 PubChem CID: 49831000 SMILES: CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC
MilliporeSigma™ Calbiochem™ InSolution™ Staurosporine, Streptomyces sp.
InSolution™ Staurosporine, Streptomyces sp. CAS 62996-74-1, is provided as a 1mm (100μg/214μl) solution of Staurosporine (Cat. No. 569397) in DMSO
R&D Systems™ Staurosporine
A Tocris product, part of R&D Systems. Staurosporine synthesized to Ancillary Material Grade
R&D Systems™ Staurosporine
CAS: 62996-74-1 Molecular Formula: C28H26N4O3
Staurosporine, Fisher BioReagents
CAS: 62996-74-1 Molecular Formula: C28H26N4O3 Molecular Weight (g/mol): 466.541 InChI Key: HKSZLNNOFSGOKW-ZYSRIHRCSA-N Synonym: staurosporine,kinome_3629 PubChem CID: 49831000 SMILES: CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC
Novus Biologicals™ Staurosporine Inhibitor
Low molecular weight, organic compound that may help regulate biological process.
Staurosporine, 98%, Thermo Scientific™
CAS: 62996-74-1 Molecular Formula: C28H26N4O3 Molecular Weight (g/mol): 466.541 InChI Key: HKSZLNNOFSGOKW-ZYSRIHRCSA-N Synonym: staurosporine,kinome_3629 PubChem CID: 49831000 SMILES: CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC