Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staurosporine, 98%, Thermo Scientific™

Catalog No. AC328530002
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328530002 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC328530002 Supplier Thermo Scientific Chemicals Supplier No. 328530002

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62996-74-1
Molecular Formula C28H26N4O3
Molecular Weight (g/mol) 466.541
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
Synonym staurosporine, kinome_3629
PubChem CID 49831000
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC

Specifications

Melting Point 270.0°C
Quantity 0.25 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in ethylacetate
Formula Weight 466.5
Chemical Name or Material Staurosporine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.