Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staurosporine, Fisher BioReagents

Catalog No. BP2541500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
Packaging:
Poly Micro Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2541500 500 μg Poly Micro Tube
BP2541100 100 μg Poly Micro Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP2541500

missing translation for 'validMainframeMsg'

Staurosporine is a potent protein kinase inhibitor useful for studies on protein phosphorylation in the regulation of cellular functions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62996-74-1
Molecular Formula C28H26N4O3
Molecular Weight (g/mol) 466.541
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
Synonym staurosporine, kinome_3629
PubChem CID 49831000
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC

Specifications

Melting Point 270°C
Color White
Assay Percent Range > 98.0 %
Packaging Poly Micro Tube
Quantity 500 μg
Merck Index 15, 8928
Residual Solvents Pass Test
Formula Weight 466.5
Percent Purity >98.0%
Physical Form Solid
Chemical Name or Material Staurosporine, From Streptomyces sp.
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
Limited evidence of a carcinogenic effect. May be harmful by inhalation, in contact with skin and if swallowed. Moisture sensitive. Sensitivity to light. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Use personal protective equipment. Do not breathe dust. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:1
Flammability:1
Instability:1
Recommended Storage 4°C, protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.