Learn More
Novus Biologicals™ Staurosporine Inhibitor

Description
Biological Activity: Broad spectrum protein kinase inhibitor. Enzymes inhibited include protein kinase C (IC50 = 3nM), protein kinase A (IC50 = 7 nM), p60v-src tyrosine protein kinase (IC50 = 6nM) and CaM kinase II (IC50 = 20nM)
Storage: Store at 4°C.
Note: Inhibitors are guranteed for 1 year from date of receipt.
Specifications
Specifications
| Color | Yellow |
| Product Type | Inhibitor |
| Target Proteins | Staurosporine |
| CAS | 62996-74-1 |
| Molecular Formula | C28H26N4O3 |
| Formula Weight | 466.44g/mol |
| Solubility | Soluble to 50mM in DMSO |
| Content And Storage | Store at 4°C |
| For Use With (Application) | Inhibits apoptosis |
| Quantity | 100 μg |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.