Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staurosporine, MP Biomedicals™

Catalog No. MP219140090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219140090 500 μg
MP219140091 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219140090 Supplier MP Biomedicals, Inc Supplier No. 0219140090
Add to Cart
Add to Cart

Possesses inhibitory activity against fungi and yeasts. Staurosporine, MP Biomedicals is an antibiotic that is isolated from Streptomyces species. It is a potent inhibitor of phospholipid/Ca2+-dependent protein kinases and is used to study the involvement

Starosporin is used to study the involvement of protein kinases in signal transduction pathways. It arrests normal cells in the G1 checkpoint and is a potent inhibitor of Protein kinase C and other protein kinases in vitro and in cultured cells. It is an inducer of apoptosis in many mammalian cell types.

  • Potent inhibitor of phospolipid/calcium dependent and cyclic nucleotide dependent protein kinases
  • Useful as a tool for studies on protein phosphorylation in the regulation of cellular functions
  • Inhibits the upregulation of VEGF expression in tumor cells
  • Partially reverses MDR, sensitizing cells with MDR phenotype to cytotoxic agents
  • Inhibits Pgp phosphorylation (functional significance of Pgp phosphorylation is ill defined)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62996-74-1
Molecular Formula C28H26N4O3
Molecular Weight (g/mol) 466.541
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
Synonym Staurosporin, Antibiotic AM-2282
PubChem CID 49831000
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC

Specifications

Physical Form Powder
Melting Point 237°C
Quantity 500 μg
Solubility Information May be reconstituted by dissolving the lyophilized reagent in DMSO (5mg/mL, clear, colorless solution), DMF, ethyl acetate (5mg/mL, clear colorless solution) or methanol or water.
Formula Weight 466.5
Percent Purity ≥98%
Recommended Storage 2-8°C, protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.