Learn More
Cancer Diagnostics, Inc.™ Stealth Safety Scalpels
SDP
Catalog No. 22500468 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Size:
# 22
# 60
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-500-468 # 60
22-500-213 # 22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-500-468 Supplier Cancer Diagnostics, Inc.™ Supplier No. FS0060
Add to Cart
Add to Cart
  • Integrated rigid plastic handle with protective sheath.
  • Slide sheath down to use, slide up when not in use.
  • Dispose when dull.
Sizes:
#22 and #60 and #70 (round tip)
Specifications
Product Type Finger Scalpel
Size # 60
Disposable Single-use
Sterility Non-sterile
Includes Handle
Handle Material Plastic