Learn More
Labconco™ SteamScrubber™ and FlaskScrubber™ Laboratory Glassware Washer Accessory, Freestanding to Mobile Conversion Kit
SDPSCI_ED
Catalog No. S28363 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
SteamScrubber and FlaskScrubber Laboratory Glassware Washer
Catalog No. For Use With (Equipment)
S28363 SteamScrubber and FlaskScrubber Laboratory Glassware Washer
1 options
Catalog No. S28363 Supplier Labconco™ Supplier No. 4596100/EMD
Add to Cart
Add to Cart
For use with SteamScrubber™ and FlaskScrubber™ Laboratory Glassware Washer. Labconco™ Freestanding to Mobile Conversion Kit add mobility to freestanding glassware washers.
  • Add mobility to freestanding SteamScrubber and FlaskScrubber glassware washer
  • Allow washers to be plugged into a 20A electrical wall outlet and connected to standard laboratory sink faucet
  • Includes four 5.08cm (2 in.) diameter rubber casters, two aerator adapters for connection to standard 0.85 in.- 27 male and 0.93 in. - 27 female laboratory faucets, drain hose, installation instructions, 3.04m (10 ft.) power cord and NEMA 5/20 plug
  • Not for use with undercounter washers or FlaskScrubber Vantage Series Glassware Washers
Specifications
Product Type Freestanding to Mobile Conversion Kit
For Use With (Equipment) SteamScrubber and FlaskScrubber Laboratory Glassware Washer
Weight (English) Shipping 60 lb.
Weight (Metric) Shipping 27.2 kg
Electrical Requirements 115V, 60Hz, 20A, NEMA 5/20