Learn More
Description
A suggested positive control is human spleen tissue lysate. PA5-20404 can be used with blocking peptide PEP-0521.
Specifications
Specifications
| Antigen | STEAP1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | STEAP1 |
| Gene Accession No. | Q9CWR7, Q9UHE8 |
| Gene Alias | 2410007B19Rik; metalloreductase STEAP1; PRSS24; six transmembrane epithelial antigen of the prostate 1; six-transmembrane epithelial antigen of prostate 1; Steap; Steap1; STEAP-1; tcag7.1170 |
| Gene Symbols | STEAP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
