Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STEAP2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176823
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176823 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176823 Supplier R&D Systems Supplier No. NBP176823

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

STEAP2 Polyclonal specifically detects STEAP2 in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen STEAP2
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 2.5 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q8NFT2
Gene Alias EC 1.16.1, EC 1.16.1.-, IPCA1, metalloreductase STEAP2, PCANAP1STMP, prostate cancer associated protein 1, Prostate cancer-associated protein 1, Protein upregulated in metastatic prostate cancer, PUMPCn, six transmembrane epithelial antigen of prostate 2, six transmembrane epithelial antigen of the prostate 2, Six-transmembrane epithelial antigen of prostate 2, SixTransMembrane protein of prostate 1, STAMP1IPCA-1
Gene Symbols STEAP2
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human STEAP2. The immunogen is located within the last 50 amino acids of STEAP2. Amino Acid Squence: KLKRIKKGWEKSQFLE
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 261729
Test Specificity This STEAP2 antibody does not cross-react with other STEAP proteins.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.