Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate 98.0+%, TCI America™
SDP

Catalog No. D164425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D164425G 25 g
D1644100G 100 g
D1644500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D164425G Supplier TCI America Supplier No. D164425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2082-79-3
Molecular Formula C35H62O3
Molecular Weight (g/mol) 530.878
MDL Number MFCD00017498
InChI Key SSDSCDGVMJFTEQ-UHFFFAOYSA-N
Synonym 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid Stearyl Ester, Octadecyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate
PubChem CID 16386
IUPAC Name octadecyl 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)CCC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

Melting Point 53°C
Color White
Quantity 25 g
Formula Weight 530.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Stearyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate
EINECSNumber (3)-1737
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.