Learn More
Description
- Manufactured from premium corrosion-resistant stainless steel for maximum durability and longevity
- Low form design provides stability and minimizes spillage during heating or mixing
- Rolled rim edge ensures easy handling and smooth pouring of liquids
- Uniform wall thickness supports even heat distribution and resistance to denting
- Non-reactive surface safe for use with most reagents and solutions
- Autoclavable and reusable, ideal for sterilized laboratory environments
- Available in capacities from 32 ml to 5000 ml to meet diverse lab needs
- Lightweight yet sturdy construction for dependable daily use
- Suitable for general lab, educational, chemical, and industrial environments
- Ideal for sample preparation, solution heating, reagent mixing, and material storage
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | Stainless Steel |
| Type | Steel Beakers |
| Graduated | No |
| Style | Griffin, Low Form |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.