Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Catalog Number | Mfr. No. | Animal | Model | Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Animal | Model | Size | Price | Quantity & Availability | |||
10-000-760
![]() |
Stoelting™
51377 |
Mouse | Coronal section | 0.5 mm |
Each for $878.50
|
|
|||
10-000-761
![]() |
Stoelting™
51378 |
Rat | Coronal section | 0.5 mm |
Each for $998.50
|
|
|||
10-000-762
![]() |
Stoelting™
51379 |
Mouse | Coronal/Sagittal section | 1.0 mm |
Each for $855.00
|
|
|||
Specifications
Spinal Cord |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title