Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Steel Inclined Plane with Car and Pulley
SCI_ED

Catalog No. S27663 Shop All Eisco Products
Change view
Click to view available options
Length (English):
48 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S27663 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27663 Supplier Eisco™ Supplier No. PH0335
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Steel Inclined Plane with Car and Pulley is 48 in. long, straight as an arrow, and can accommodate any type of car

  • Made of heavy steel with rounded corners
  • Holes in the supporting flanges fit a 12 mm support rod
  • Includes car, clamp pulley, string, and basket
  • Stand base, rod, and support clamp are sold separately

Specifications

Product Type Inclined Plane
Description Rounded Corners
Material Steel
For Use With (Application) Motion and Energy Demonstartions
For Use With (Equipment) 12 mm Support Rod
Includes Basket, Car, Clamp Pulley, and String
Length (English) 48 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.