Learn More
Newcomer Supply Steiner-Chapman Modified Silver Stain Kit, 500 ml kit - No Uranyl Nitrate!
SDP
Supplier:  Newcomer Supply 9172B
Encompass
Newcomer Supply Steiner-Chapman Modified Silver Stain Kit procedure, with included microwave modification, is used for staining spirochetes, Helicobacter pylori, Legionella pneumophila, other nonfilamentous bacteria and fungus. This modified method eliminates the use of Uranyl Nitrate and its regulatory and disposal requirements.
Catalog No. NC2102293
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.