Learn More
Neuromics, Inc Stem Cell Complete Low Serum Media, 500 ml is a low serum complete medium designed for the culture of cells. The medium consists of basal medium supplemented with 2% FBS.
Supplier:  Neuromics, Inc CAFM01
Encompass
Stem Cell Complete Low Serum Media (SCLSM) is a low serum complete medium designed for the culture cells. It was tested and optimized with cell growth and proliferation in vitro. It is formulated for use with 5% CO2 and 95% air in a humidified incubator.The medium consists of 500 ml of basal medium (containing essential and non-essential amino acids, vitamins, organic and inorganic compounds, hormones, growth factors, trace minerals), supplemented with 2% fetal bovine serum.SCLSM are subjected to comprehensive quality control tests using primary human cells. Each lot is checked for growth-promoting activity, adherence rate, and typical morphology of the tested cells. Approved in-house lots of variety of human mesenchymal stem cells are used as a reference. In addition, every lot of media is tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Catalog No. NC2064300
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.