Learn More
Enzo Life Sciences Stem cell factor (human), (recombinant) (2µg)
SDP
Supplier:  Enzo Life Sciences ALX2018040002
Encompass_Preferred
Alternative name: SCF, c-Kit ligand, KL, Steel factor, MGF. Endotoxin: ≤1 EU/μg protein measured by kinetic LAL analysis. Purity: ≥98% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm). MW: ~18.4kDa. Appearance: White lyophilized (freeze-dried) powder. Formulation: Lyophilized from 10mM acetic acid (AcOH) at a concentration of 1mg/ml. Sterile filtered. Source: Produced in E. coli. Non-glycosylated protein, containing 165 amino acids. Long Term Storage: -20°C. Use/Stability: It is recommended that a carrier protein (0.1% HSA or BSA) is added for long term storage. Handling: Centrifuge the vial before opening the cap. After reconstitution, prepare aliquots and store at -20°C.
Catalog No. 50-200-8001
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.