Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Engino™ STEM Challenge Bundle, Stem Mechanics and Newton's Law
SDPSCI_ED

Catalog No. S00027 Shop All United Scientific Supplies, Inc Products
Click to view available options
Grade:
4 to 12
Choking Hazard
Choking Hazard

Includes two Engino Kits that are ideal for STEM Challenge Competitions.

  • Includes two Engino Kits:
  • STEM901: STEM Mechanics: Levers, Linkages and Structures.
  • Includes a manual with instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
  • STEM07: STEM, Newton's Laws: Inertia, Momentum, Kinetic, and Potential Energy.
  • Includes a manual with instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different model

Specifications

Grade 4 to 12
Format Kit
Product Type STEM Challenge Kit
Languages English
Subject Science
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.