Learn More
Walter Products™ Stem (Dicotyledon) Column Model
SCI_ED
Catalog No. S139172 Shop All Walter Products Products
Change view
Click to view available options
Dimensions (L x W x H):
12 x 11.8 x 12.6 in.
Catalog No. Dimensions (L x W x H)
S139172 12 x 11.8 x 12.6 in.
1 options
This item is not returnable. View return policy
Catalog No. S139172 Supplier Walter Products™ Supplier No. B10304
Add to Cart
Add to Cart
This item is not returnable. View return policy
This 120 times enlarged model highlights the details of a dicotyledonous stem in vivid colors.
  • Shows the longitudinal and cross section of the periderm, cortex, primary phloem, phloem ray, cambium, annual ring, wood ray, primary xylem, and marrow
  • Mounted on a base
Specifications
Dimensions (L x W x H) 12 x 11.8 x 12.6 in.
Material PVC
Product Type Anatomical Model