Learn More
Beckman Coulter Stem-Kit Reagents, 50 Tests, CE, IVD
Supplier:  Beckman Coulter IM3630
Encompass_Preferred
Stem-Kit Reagents is intended for the simultaneous identification and enumeration of CD45+ and dual-positive CD45+ CD34+ cell population percentages and absolute counts in biological specimens by flow cytometry. Biological specimens include fresh normal or mobilized peripheral whole blood, and fresh or thawed apheresis products, cord blood and bone marrow. Cell population measurements may be obtained using either an automated method or a manual method for gating and analysis.
Catalog No. NC0896707