Learn More
Walter Products™ Stem (Monocotyledon) Model
SCI_ED
Catalog No. S139171 Shop All Walter Products Products
Change view
Click to view available options
Dimensions (L x W x H):
16.5 x 17 x 6 in.
Catalog No. Dimensions (L x W x H)
S139171 16.5 x 17 x 6 in.
1 options
This item is not returnable. View return policy
Catalog No. S139171 Supplier Walter Products™ Supplier No. B10302
Add to Cart
Add to Cart
This item is not returnable. View return policy
This model of corn gives details of cellar anatomy of the generalized monocot in cross and longitudinal section.
  • Shows important structures such as cuticular layer, epidermis, thick walled mechanical tissue, fundamental parenchymatous tissue and the details of closed vascular bundles
  • Enlarged 45 times and made from high quality PVC
Specifications
Dimensions (L x W x H) 16.5 x 17 x 6 in.
Material PVC
Product Type Anatomical Model