Learn More
Stemculture LLC StemCultures LLC StemBeads Blank15
SDP
Supplier:  Stemculture LLC SB001
Encompass
StemBeads Blank15 is a microparticle containing all typical components of StemBeads products except the protein. StemBeads Blank15 are microbeads composed of an FDA approved, biodegradable PLGA-based polymer. The nature of StemBeads Blank15 is to serve as a control for your experiments.
Catalog No. NC2670066
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.