Learn More
Amsbio LLC StemFit Basic03 (AOF, defined medium for human pluripotent stem cell culture)
Supplier:  Amsbio LLC SFB503
Encompass_Preferred
StemFit is a range of animal-origin free, chemically defined media proven to effectively maintain induced pluripotent stem cells (iPSCs) and embryonic stem cells (ESCs) under feeder-free conditions during the reprogramming, expansion and differentiation phases of stem cell culture and growth medium. StemFit Basic03 MAIN FEATURES: Suitable for clinical research| Superior expansion with single-cell passaging | Chemically defined formula minimizes lot-to-lot variation | Consistent high performance and compatibility with various matrices | Animal-Origin-Free & feeder-free | Weekend-free cell culture | Reduced media consumption | Reproducible growth rates | Easy transition from feeder cells | Superior colony forming efficiency from a single clone.
Catalog No. NC1623494
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.