Learn More
Description
Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into adipocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.
- Contains high quality differentiation factors to drive reproducible and efficient MSC adipogenesis
- Is defined to reduce unwanted experimental variability
- Has been developed and optimized using MSCs
Specifications
Specifications
| Product Line | R&D Systems |
| Quantity | 3.5 mL |
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
