Learn More
R&D Systems™ StemXVivo™ Cardiomyocyte Differentiation Kit
SDP
Catalog No. SC032B Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SC032B 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. SC032B Supplier R&D Systems™ Supplier No. SC032B
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Includes
Stem Cell Qualified RGF BME, Pathclear, Cardiomyocyte Differentiation Base Media Supplement I, Cardiomyocyte Differentiation Base Media Supplement II, Cardiomyocyte Differentiation Cocktail I, Cardiomyocyte Differentiation Cocktail IIA, Cardiomyocyte Diff
Used for drug toxicity and small molecule screening.
  • Optimized reagents generate more strong and uniform beating of differentiated cardiomyocytes
  • Provides consistent differentiation across pluripotent stem cell lines
  • Reproducible differentiation protocols translate into cost and time savings
  • Maximizes workflow efficiency by standardizing cardiomyocyte differentiation
Specifications
Product Type Stem Cell Differentiation Media
Quantity 1 Kit
Content And Storage Stem Cell Qualified RGF BME - Pathclear, Cardiomyocyte Differentiation Base Media Supplement I, Cardiomyocyte Differentiation Base Media Supplement II, Cardiomyocyte Differentiation Cocktail I, Cardiomyocyte Differentiation Cocktail II, Cardiomyocyte Differentiation Cocktail III, Anti-Human Cardiac Troponin T Antibody
Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
For Laboratory Research Use Only. Not for Diagnostic use.